missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ Butterfield's Phosphate Buffer (pH 7.2 stock solution)

Catalog No. R23700
Change view
Click to view available options
Capacity (Metric):
90 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
R23700 90 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R23700 Supplier Thermo Scientific™ Supplier No. RE23700

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Formulation used in food, dairy and pharmaceutical testing by AOAC, APHA, FDA, USDA, and USP test methods.

Thermo Scientific™ Remel Butterfield's 90mL Phosphate Buffer (pH 7.2 stock solution) formulation is used in food, dairy and pharmaceutical testing as referenced in AOAC, APHA, FDA, USDA, and USP test methods.

  • Dilution Bottles are leakproof, wide mouth PET plastic with tear away sterility strip and easy, one-hand opening hinged lid
  • Gamma irradiated
  • Color-coded (green) cap for easy and accurate visual identification

Specifications

Capacity (Metric) 90 mL
Color Clear
Description Butterfield's; 90mL
For Use With (Application) AOAC, APHA, FDA, USDA, UPSP food, dairy, pharmaceutical testing methods
Material PET
Sterility Sterile
Format Green Cap - 90mL/Bottle
Graduated No
Mouth Wide
Closure Material Polyethylene
Closure Type Hinged Lid
Through the Closure Dispensing No
Shape Round
Certifications/Compliance AOAC; APHA; FDA; USDA; USP
Product Type Dilution Bottle
Show More Show Less