missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ BactiDrop™ Nitrate B

Catalog No. R21538
Change view
Click to view available options
Quantity:
50 x 0.75 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R21538 50 x 0.75 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R21538 Supplier Thermo Scientific™ Supplier No. RE21538

Determine the ability of an organism to reduce nitrate to nitrite or free nitrogen gas using Nitrate Reagent B.

Determine the ability of an organism to reduce nitrate to nitrite or free nitrogen gas using Thermo Scientific™ Remel™ BactiDrop™ Nitrate Reagent B. This ability is characteristic of the Enterobacteriaceae1 family. Nonfermenters and other miscellaneous gram negative bacilli vary in their ability to reduce nitrates, with some being capable of further reducing nitrite to nitrogen gas2. In 1962, Daubner reported that all members of the Enterobactericeae tested, with the exception of Erwinia species, reduced nitrate in eight hours3.

Nitrate serves as a source of nitrogen for many bacteria and fungi, but in order to be utilized it must be broken down.Ready to use: Convenience of a prepared reagent.Ease of Differentiation: Positive test indicates red color development after addition of reagents; and no color development after addition of zinc dust. Negative test indicates no color development after addition of reagents and red color development after addition of zinc dust.Rapid Detection: Immediate results.The nitrite reduction test is a two-step procedure. Sulphanilic acid is added to acidify the medium and then naphthylamine is added which reacts with the nitrites to produce a red color. Certain bacteria convert nitrate to nitrite, ammonia, or gas. This process is denitrification. If no color develops after addition of reagents, the organism has either reduced the nitrate to free nitrogen or it has not attacked the nitrate at all. Nitrate will react with zinc dust to form nitrite. A pink to red color development after addition of zinc dust indicates that the nitrates are present, indicating a negative nitrite reduction test. No formation of a red color after addition of zinc dust indicates that the organism has reduced the nitrate to free nitrogen or ammonia and is considered a positive nitrate test2.Used in conjunction with Nitrate Reagent A, Part No. R21536.Not all products are available for sale in all territories. Please inquire.Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

Specifications

Content And Storage 20°C to 25°C
Description BactiDrop Nitrate B
CE Marker Yes
Quantity 50 x 0.75 mL