Learn More
Thermo Scientific™ Remel™ BactiCard™ Candida Kit
Catalog No. R21106
Change view
Click to view available options
Quantity:
25 Tests/Kit
Catalog No. Quantity
R21106 25 Tests/Kit
1 options
Catalog No. R21106 Supplier Thermo Scientific™ Supplier No. RE21106
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Thermo Scientific™ Remel BactiCard Candida Kit is used for presumptive identification of Candida albicans using enzyme technology.
Thermo Scientific™ Remel BactiCard Candida Kit is used for presumptive identification of Candida albicans using enzyme technology.
  • For use in qualitative procedures for the presumptive identification of Candida albicans by the rapid detection of L-Proline aminopeptidase and Β-galactosaminidase
  • Test results in 30 seconds
  • 4-Methylumbelliferyl-N-acetyl-Β-D-galactosaminide in the presence of the enzyme Β-galactosaminidase releases 4-methylumbelliferone, which is a highly fluorescent compound detectable by ultraviolet light
  • L-Proline aminopeptidase hydrolyzes L-Proline-Β-napthylamide and reacts with the color developer to form a red color If PRO and MUGAL are both positive, then C. albicans is present Product is ready for use and no further preparation is necessary
Includes:
Test cards, rehydrating fluid, color developer, MUGAL reagent and applicator sticks.
Specifications
Content And Storage Store in its original container at 2° to 8°C until used.
Description BactiCard™ Candida
Quantity 25 Tests/Kit
For in vitro diagnostic use