Learn More
Stemgent™ Alvetex™ Perfusion Plates with Alvetex Scaffold Well Inserts
Dynamic circulation and perfusion of culture medium within a multi-welled plate
$1565.57 - $3193.97
Specifications
| Material | Polystyrene |
|---|---|
| For Use With (Application) | Cell culture laboratory |
| Surface Treatment | Non-tissue Culture Treated |
| Sterility | Sterile |
| Catalog Number | Mfr. No. | No. of Wells | Inserts per Plate | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | No. of Wells | Inserts per Plate | Price | Quantity | ||||
AVPKIT3
|
Stemgent™
AVPKIT3 |
6 | 12 |
Pack for $1,759.83
|
|
||||
AVPKIT4
|
Stemgent™
AVPKIT4 |
12 | 12 |
Pack for $1,565.57
|
|
||||
AVPKIT5
|
Stemgent™
AVPKIT5 |
6 | 48 |
Pack for $3,193.97
|
|
||||
AVPKIT6
|
Stemgent™
AVPKIT6 |
12 | 48 |
Pack for $3,193.97
|
|
||||
Description
- Manufactured from non-tissue culture-treated polystyrene
- Sterile, supplied in blister pack
- With luer locks
- Plate includes lid and Leur-lock fittings for tube attachment
- Designed for use in cell culture laboratory
- Available as 6- and 12-well inserts
Specifications
| Polystyrene | |
| Cell culture laboratory | |
| Non-tissue Culture Treated | |
| Sterile |