Learn More
Stemgent™ Alvetex™ Perfusion Plates with Alvetex Scaffold Well Inserts
Dynamic circulation and perfusion of culture medium within a multi-welled plate
Supplier: Stemgent™ AVPKIT6
Description
- Manufactured from non-tissue culture-treated polystyrene
- Sterile, supplied in blister pack
- With luer locks
- Plate includes lid and Leur-lock fittings for tube attachment
- Designed for use in cell culture laboratory
- Available as 6- and 12-well inserts
Specifications
| 48 | |
| Cell culture laboratory | |
| Non-tissue Culture Treated | |
| 12 |
| Polystyrene | |
| 5 perfusion plates w/48 x Alvetex Scaffold 12 well inserts | |
| Sterile |