Learn More
Stemgent™ Alvetex™ Perfusion Plates with Alvetex Scaffold Well Inserts DFS Item
Dynamic circulation and perfusion of culture medium within a multi-welled plate
Supplier:  Stemgent™ AVPKIT3
 View more versions of this product
Catalog No. AVPKIT3
Only null left
Explore available promotions
Description
Description
  • Manufactured from non-tissue culture-treated polystyrene
  • Sterile, supplied in blister pack
  • With luer locks
  • Plate includes lid and Leur-lock fittings for tube attachment
  • Designed for use in cell culture laboratory
  • Available as 6- and 12-well inserts
Specifications
Specifications
12
Cell culture laboratory
Non-tissue Culture Treated
6
Polystyrene
2 perfusion plates w/12 x Alvetex Scaffold 6 well inserts
Sterile
Product Suggestions
Product Suggestions