missing translation for 'onlineSavingsMsg'
Learn More

Reference Material for Flash Point Certified by The Japan Petroleum Institute, Hexadecane, TCI America™

Catalog No. S0555250ML
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0555250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S0555250ML Supplier TCI America Supplier No. S0555250ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 544-76-3
Molecular Formula C16H34
Molecular Weight (g/mol) 226.448
InChI Key DCAYPVUWAIABOU-UHFFFAOYSA-N
Synonym n-hexadecane, cetane, n-cetane, hexadecan, hexadekan, cetan, zetan, unii-f8z00shp6q, ccris 5833, pentadecane, methyl
PubChem CID 11006
ChEBI CHEBI:45296
IUPAC Name hexadecane
SMILES CCCCCCCCCCCCCCCC

Specifications

Color Colorless
Quantity 250 mL
Formula Weight 226.45
Physical Form Liquid
Chemical Name or Material Reference Material for Flash Point Certified by The Japan Petroleum Institute, Hexadecane
EINECSNumber (2)-0010
RTECSNumber ML9200000
TSCA Yes