missing translation for 'onlineSavingsMsg'
Learn More

Reference Material for Flash Point Certified by The Japan Petroleum Institute, Decane, TCI America™

Catalog No. S0554250ML
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0554250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S0554250ML Supplier TCI America Supplier No. S0554250ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 124-18-5
Molecular Formula C10H22
Molecular Weight (g/mol) 142.286
MDL Number MFCD00008954
InChI Key DIOQZVSQGTUSAI-UHFFFAOYSA-N
Synonym n-decane, nonane, methyl, decyl hydride, unii-nk85062oiy, ccris 653, hsdb 63, dsstox_cid_4913, decane, analytical standard, dsstox_rid_77577, dsstox_gsid_24913
PubChem CID 15600
ChEBI CHEBI:41808
IUPAC Name decane
SMILES CCCCCCCCCC

Specifications

Color Colorless
Quantity 250 mL
UN Number 2247
Formula Weight 142.29
Physical Form Liquid
Chemical Name or Material Reference Material for Flash Point Certified by The Japan Petroleum Institute, Decane
EINECSNumber (2)-0010
RTECSNumber HD6550000
TSCA Yes