Learn More
Thermo Scientific™ Reference Genomic DNA 103
Description
Reference Genomic DNA 103 is used as a positive control in assays.
Specifications
Specifications
| Content And Storage | Store at –25°C to –15°C |
| Quantity | 2 reactions |
Frequently Asked Questions (FAQs)
Does Ref103 come in the CytoScan XON Assay Kit (Cat. No. 931311)?
Reference Genomic DNA 103 is not included in the kit, but is available for purchase separately (Cat. No. 901012).
Note: A positive control is recommended for the CytoScan XON assay.
For Research Use Only. Not for use in diagnostic procedures.