missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Red Clickmaster Ratchet Nut for FastPrep-96™
Description
- The FastPrep™ is the only available homogenizer with multiple interchangeable adapters which allows a unique versatility in sample number, size, and temperature conditions.
- All optional adapters stand stable on the benchtop and are commonly used as tube racks for sample storage at -20°C or -80°C.
- The Securing Knob is important for maintaining the tube holder adapters on the central axis of the FastPrep-96™.
- The Red Clickmaster Ratchet Nut supports up to 6 different adapters. It is designed to catered to large sample volumes adapters like the BigFlex, TeenFlex, and more. Thanks to its high torque, it is particularly recommended for the heaviest adapters and especially for metal tube holders.
Specifications
Specifications
| Product Type | Ratchet Nut |
| Blade Material | Stainless Steel |
| Closure Color | Red |
| For Use With (Equipment) | FastPrep-96 |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.