Learn More
Alkali Scientific Rectangle Cover Glass, Cover Slip, for Microscope Slides, 24 X 50mm, No. 1.0 Thickness 0.13 to 0.17mm, 1 oz 100/Sleeve
Numéro de catalogue. 502389051
Encompass
ASI™ Rectangle Cover Glass are high quality imported glass, and are non-cytotoxic. They are manufactured in 100,000 grade clean room and resistant to organic solvents and autoclaved by high temperature.
Numéro de catalogue. 502389051 Fournisseur Alkali Scientific Code fournisseur SM2450
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
ASI™ Rectangle Cover Glass are high quality imported glass, and are non-cytotoxic. They are manufactured in 100,000 grade clean room and resistant to organic solvents and autoclaved by high temperature.