LGC Genomics Llc (Lucigen) Recovery Medium, 8 x 12 mLRecommended medium for all competent cell transformations.
Numéro de catalogue. NC9996840
Recovery Medium, 8 x 12 mLRecommended medium for all competent cell transformations.Enables the highest possible transformation efficiency.Use in the outgrowth stage of competent cell transformation to attain up to 10-folder higher transformation efficiency than with LB or SOC media.Trusted formulation used for cloning with Biosearch Technologies competent cells.
Numéro de catalogue.
NC9996840
Fournisseur
LGC Genomics LlcCode fournisseur
800261
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Recovery Medium, 8 x 12 mLRecommended medium for all competent cell transformations.Enables the highest possible transformation efficiency.Use in the outgrowth stage of competent cell transformation to attain up to 10-folder higher transformation efficiency than with LB or SOC media.Trusted formulation used for cloning with Biosearch Technologies competent cells.