missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novex™ Recombinant Protein G - Sepharose™ 4B
Description
Recombinant protein G is produced in a strain of E. coli which contains the gene for protein G from Streptococcus sp. This product is prepared by covalently coupling highly purified rec-Protein G to Sepharose™ 4B. It is supplied as a 50% suspension containing rec-Protein G-Sepharose™ 4B + PBS (10 mM phosphate-buffered saline, pH 7.4, containing 0.1% Proclin).
Applications: Immunoglobulin affinity purification and Immunoprecipitation.
Recombinant protein G binds to the Fc region of many types of immunoglobulin G including:
all four subclasses of human IgG
mouse IgG
rabbit IgG
guinea pig IgG
rat IgG
goat IgG
sheep IgG
cow IgG
horse IgG
Recombinant protein G does not bind to IgG from chicken or cat, or to human IgA, IgM, or serum albumin. It binds weakly to IgG from dog.
Applications: Immunoglobulin affinity purification and Immunoprecipitation.
Recombinant protein G binds to the Fc region of many types of immunoglobulin G including:
Recombinant protein G does not bind to IgG from chicken or cat, or to human IgA, IgM, or serum albumin. It binds weakly to IgG from dog.
Spécifications
Spécifications
| Product Type | Protein G Sepharose |
| Product Line | Sepharose |
| Column Type | Affinity |
| Stationary Phase | Protein G |
| Content And Storage | This product is supplied as a 50% suspension (2 ml Recombinant Protein G-Sepharose™ 4B + 2 ml PBS) Wet Ice |
| Quantity | 2 mL |
| Form | Suspension |
For Research Use Only. Not for use in diagnostic procedures.