missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Recombinant Protein G-HRP

Catalog No. 101223
Encompass
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
101223 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 101223 Supplier Invitrogen™ Supplier No. 101223

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Recombinant Protein G-HRP

Recombinant protein G is produced in a strain of E. coli which contains the gene for protein G from Streptococcus sp. This protein G is conjugated with highly purified horseradish peroxidase (RZ > 3.0) and is in enyme conjugate stabilizer containing 0.1% proclin.

Recombinant protein G binds to the Fc region of many types of immunoglobulin G including:

  • all four subclasses of human IgG
  • mouse IgG
  • rabbit IgG
  • guinea pig IgG
  • rat IgG
  • goat IgG
  • sheep IgG
  • cow IgG
  • horse IgG

    Recombinant protein G does not bind to IgG from chicken or cat, or to human IgA, IgM, or serum albumin. It binds weakly to IgG from dog.
  • Specifications

    Conjugate HRP
    Quantity 1 mL
    Form Liquid
    Protein Subtype Protein G
    Content And Storage 2-8°C
    Shipping Condition Wet Ice

    For Research Use Only. Not for use in diagnostic procedures.