missing translation for 'onlineSavingsMsg'
Learn More

Novex™ Recombinant Protein G

Catalog No. 101200
Encompass
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
101200 1 mg
101201 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 101200 Supplier Novex™ Supplier No. 101200

Recombinant Protein G

Recombinant protein G is produced in a strain of E. coli which contains the gene for protein G from Streptococcus sp. It is supplied as a salt-free lyophilized powder.

Recombinant protein G binds to the Fc region of many types of immunoglobulin G including:

  • all four subclasses of human IgG
  • mouse IgG
  • rabbit IgG
  • guinea pig IgG
  • rat IgG
  • goat IgG
  • sheep IgG
  • cow IgG
  • horse IgG

    Recombinant protein G does not bind to IgG from chicken or cat, or to human IgA, IgM, or serum albumin. It binds weakly to IgG from dog.
  • Specifications

    Conjugate Unconjugated
    Quantity 1 mg
    Form Powder
    Protein Subtype Protein G
    Content And Storage 2-8°C
    Shipping Condition Wet Ice

    For Research Use Only. Not for use in diagnostic procedures.