Recombinant Protein;1MG;Human DNAM-1, His Tag (DN1-H52H6) is expressed from human 293 cells (HEK293). It contains AA Glu 19 - Asn 247 (Accession # NP_006557).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 27.9 kDa. The protein migrates as 36-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human DNAM-1, His Tag (Cat. No. DN1-H52H6) at 2 g/mL (100 L/well) can bind Human CD155, Fc Tag (Cat. No. CD5-H5251) with a linear range of 3-51 ng/mL (QC tested).;FACS assay shows that Human DNAM-1, His Tag (Cat. No. DN1-H52H6) can bind to 293T cell overexpressing human CD155. The concentration of DNAM-1 used is 3 g/mL (Routinely tested).