missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Mouse RKIP/PBP His Protein

Numéro de catalogue. NBP25217401
Modifier l'affichage
Click to view available options
Quantity:
0.1 mg
0.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
NBP25217401 0.1 mg
NBP25217405 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. NBP25217401 Fournisseur Novus Biologicals™ Code fournisseur NBP2521740.1MG

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-187 of Mouse RKIP/PBP The Recombinant Mouse RKIP/PBP Protein is derived from E. coli. The Recombinant Mouse RKIP/PBP Protein has been validated for the following applications: SDS-Page.

Spécifications

Concentration 1mg/mL
For Use With (Application) SDS-PAGE
Formulation Phosphate buffered saline (pH7.4) containing 10% glycerol
Gene ID (Entrez) 5037
Molecular Weight (g/mol) TMW: 23.2kDa
Quantity 0.1 mg
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Alias HCNP, hippocampal cholinergic neurostimulating peptide, Neuropolypeptide h3, PBPPEBP-1, PEBPHCNPpp, phosphatidylethanolamine binding protein 1, phosphatidylethanolamine-binding protein 1, prostatic binding protein, Prostatic-binding protein, Raf kinase inhibitory protein, RKIPRaf kinase inhibitor protein
Common Name RKIP/PBP
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain
Afficher plus de résultats Afficher moins de résultats

For research use only and is not approved for use in humans or in clinical diagnosis