Learn More
Abcam Inc Recombinant Macaca fascicularis ENPP3/B10 protein - Active
Numéro de catalogue. 502699191
Encompass
Recombinant Macaca fascicularis ENPP3/B10 protein - Active is a Fragment protein, in the 46 to 874 aa range, expressed in Mammalian, with >95% purity, < 1 EU/g endotoxin level and suitable for SDS-PAGE, FuncS.
The product is subject to the following: Abcam Restricted Use Statement
Numéro de catalogue. 502699191 Fournisseur Abcam Inc Code fournisseur AB314536100UG
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Recombinant Macaca fascicularis ENPP3/B10 protein - Active is a Fragment protein, in the 46 to 874 aa range, expressed in Mammalian, with >95% purity, < 1 EU/g endotoxin level and suitable for SDS-PAGE, FuncS.
The product is subject to the following: Abcam Restricted Use Statement