missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rebamipide 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 290°C |
| Color | White |
| Quantity | 5 g |
| Formula Weight | 370.79 |
| Percent Purity | ≥98.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Rebamipide |
Safety and Handling
| RTECSNumber | VC2518500 |
| TSCA | No |