missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ PYREXPLUS™ Reagent Bottles with Hollow standard taper Stoppers
Feature special coating for added safety
$197.92 - $2,199.58
Specifications
| Coating | Pyrexplus |
|---|---|
| Material | Borosilicate Glass |
| Sterility | Non-sterile |
| Mouth | Narrow |
| Shape | Cylindrical |
Alert:
Do not place over direct heat or flame.
| missing translation for 'productCode' | missing translation for 'brand' | Capacity (Metric) | Stopper Number | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| missing translation for 'productCode' | missing translation for 'brand' | Capacity (Metric) | Stopper Number | Price | Quantity & Availability | ||||
|
028863
|
Corning™
61500125 |
125 mL | 19 |
Case of 12 Each for $2,199.58
|
|
||||
|
028864
|
Corning™
61500250 |
250 mL | 19 |
Each for $197.92
Case of 6 Each for $1,187.52
|
|
||||
|
028865
|
Corning™
61500500 |
500 mL | 24 |
Case of 6 Each for $1,314.68
|
|
||||
|
028866
|
Corning™
615001L |
1 L | 29 |
Case of 6 Each for $1,927.84
|
|
||||
|
028867
|
Corning™
615002L |
2000 mL | 29 |
Case for $695.46
|
|
||||
Description
Features a protective PVC coating for longer product life and safety. Protective coating helps prevent glass from shattering and reduces spills. Autoclavable (121°C) and resistant to thermal shock. Bottle has a ground neck with a standard taper stopper.
For stopper only, see Cat. No. 7575.
Warning: Do not place over direct heat or flame. Do not heat above 121°C moist heat or 110°C dry heat.
Not recommended for dry heat sterilization. Will Melt.
Specifications
| Pyrexplus | |
| Non-sterile | |
| Cylindrical |
| Borosilicate Glass | |
| Narrow |