missing translation for 'onlineSavingsMsg'
Learn More

Reagent Alcohol (Denatured Alcohol), 70% (v/v), Ricca Chemical

Catalog No. 25467032
Change view
Click to view available options
Quantity:
1 L
4 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
Safety-coated Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
25467032 1 L Natural Poly Bottle
2546701SC 4 L Safety-coated Glass Bottle
25467025 10 L Cubitainer
2546705 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 25467032 Supplier Ricca Chemical Company Supplier No. 2546.7032
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Reagent Alcohol (Denatured Alcohol), 70% (v/v)

  • Reagent Alcohol (Denatured Alcohol), 70% (v/v)

Chemical Identifiers

CAS 64-17-5 , 7732-18-5 , 67-63-0 , 67-56-1
Molecular Weight (g/mol) Mixture
PubChem CID 702
ChEBI CHEBI:16236

Specifications

Melting Point -50°C
Color Colorless
CAS Min % 62.0, 29.4, 3.43, 3.09
CAS Max % 65.0, 30.6, 3.57, 3.21
Quantity 1 L
Flash Point 13°C (55.47°F)
Packaging Natural Poly Bottle
Physical Form Liquid
Molecular Formula C2H6O
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
Boiling Point 93°C
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
SMILES CCO
IUPAC Name ethanol
Percent Purity 70%
Grade Laboratory
Concentration or Composition (by Analyte or Components) 70% (v/v)
Chemical Name or Material Alcohol, Reagent, Denatured Alcohol
Show More Show Less