missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Reagent Alcohol (Denatured Alcohol), 70% (v/v), Ricca Chemical
Description
- Reagent Alcohol (Denatured Alcohol), 70% (v/v)
Specifications
Specifications
| Melting Point | -50°C |
| Color | Colorless |
| CAS Min % | 62.0, 29.4, 3.43, 3.09 |
| CAS Max % | 65.0, 30.6, 3.57, 3.21 |
| Quantity | 4 L |
| Flash Point | 13°C (55.47°F) |
| Packaging | Safety-coated Glass Bottle |
| Physical Form | Liquid |
| Molecular Formula | C2H6O |
| Synonym | ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol |
| Show More |