missing translation for 'onlineSavingsMsg'
Learn More

Reagent Alcohol, Certified, 70% (v/v), LabChem™

Catalog No. p-5379511
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
Packaging:
Cubitainer
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC222102 1 L Poly Bottle
LC222104 4 L Poly Bottle
LC222105 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC222102 Supplier LabChem Supplier No. LC222102
Only null left

Chemical Identifiers

CAS 64-17-5 , 7732-18-5 , 67-63-0 , 67-56-1
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.069
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

Melting Point -115°C
Color Colorless
CAS Max % 56.46, 37.63, 3.11, 2.8
Decomposition Information Fume; Carbon monoxide; Carbon dioxide; May release flammable gases
Density 0.881g/mL
Quantity 1 L
Flash Point 25°C
Identification Passes Test
Packaging Poly Bottle
Physical Form Liquid
Specific Gravity 0.881g/L ±0.008
Linear Formula C2H6O
UN Number UN1987
Boiling Point 78°C
Solubility Information Soluble in water
Formula Weight 46.07
Percent Purity 70% (v/v)
Grade Certified
Chemical Name or Material Reagent Alcohol
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Suspected of damaging fertility or the unborn child.
Causes damage to organs (central nervous system, optic nerve) (oral, dermal).
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep away from heat, hot surfaces, open flames, sparks.
No smoking.
Keep container tightly closed.
Keep cool.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, lighting, ventilating equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist, spray, vapors.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
Wear eye protection, face protection, protective clothing, protective gloves.
Wash exposed skin thoroughly after handling.
If exposed or concerned: Get medical advice/attention.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
If skin irritation occurs: Get medical advice/attention.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
In case of fire: Use carbon dioxide (CO2), powder, alcohol-resistant foam to extinguish.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.
EINECSNumber 200-578-6
Recommended Storage Room Temperature