missing translation for 'onlineSavingsMsg'
Learn More

Reagent Alcohol 70% (140 Proof), Reagents

Catalog No. 01337540
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337540 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01337540 Supplier Labchem Inc Supplier No. C21204504A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Reagent Alcohol 70% (140 Proof), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 64-17-5 , 67-56-1 , 7732-18-5 , 67-63-0
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
IUPAC Name ethanol
SMILES CCO

Specifications

Color Colorless
Quantity 4 L
Packaging Poly Bottle
Physical Form Liquid
Specific Gravity 0.885
Percent Purity 70%
Grade Reagent
Chemical Name or Material Reagent Alcohol 70% (140 Proof)