missing translation for 'onlineSavingsMsg'
Learn More

Reagent Alcohol, 50% (v/v), Ricca Chemical

Catalog No. p-3252213
Change view
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
25465016 500 mL Natural Poly Bottle
2546501 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 25465016 Supplier Ricca Chemical Company Supplier No. 2546.5016
Only null left

Chemical Identifiers

CAS 7732-18-5 , 64-17-5 , 67-63-0 , 67-56-1
Molecular Formula C2H6O
Molecular Weight (g/mol) Mixture
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

Color Colorless
CAS Min % 49.0, 45.0, 2.0, 2.0
CAS Max % 51.0, 46.0, 3.0, 3.0
Quantity 500 mL
Packaging Natural Poly Bottle
Physical Form Liquid
Boiling Point 85°C
Percent Purity 50%
Grade Laboratory
Concentration or Composition (by Analyte or Components) 50% (v/v)
Chemical Name or Material Alcohol, Reagent
Show More Show Less