missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ReadyProbes™ 2.5% Normal Goat Serum (1X)

Catalog No. R37624
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R37624 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R37624 Supplier Invitrogen™ Supplier No. R37624

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ready-to-use non-specific blocker for use in immunohistochemistry (IHC), immunocytochemistry (ICC), and immunofluorescence (IF) experiments

  • ReadyProbes 2.5% Normal Goat Serum is a ready-to-use non-specific blocker for use in immunohistochemistry (IHC), immunocytochemistry (ICC), and immunofluorescence (IF) experiments, usually when a goat secondary antibody is used.
  • Incubating cells/tissue with diluted serum from the same species as the host of the secondary antibody typically reduces non-specific binding.
  • Simply cover the tissue section or cells with a few drops of ReadyProbes 2.5% Normal Goat Serum and incubate for 30–60 minutes at room temperature.
  • Ready Probes 2.5% Normal Goat Serum can also be used to dilute primary and secondary antibodies.
  • Ready Probes 2.5% Normal Goat Serum is provided as a ready-to-use reagent with no further dilution or preparation required.
  • The serum is collected from healthy adult animals and the heat-treated, centrifuged, and filtered. It is then checked for cross reactivity using an appropriate immunoassay.
  • The serum is diluted to 2.5% (v/v) in buffer, with 0.08% sodium azide added as a preservative.

Specifications

Content And Storage Provided as 50-mL solution at ready-to-use (1X) concentration in a clear plastic dropper bottle.

Store at 2–8°C.

When stored as directed, product can be used for 6 months after receipt.

Species Goat
Form Liquid
Product Type Goat Serum
Quantity 50 mL
Product Line ReadyProbes
Reagent Type Blocking Reagent
Shipping Condition Room Temperature or Wet Ice
Can I use ReadyProbes 2.5% Normal Goat Serum (1X) as an antibody diluent for both primary and secondary antibodies?

Yes, ReadyProbes 2.5% Normal Goat Serum (1X) can be used as an antibody diluent for both primary and secondary antibodies, except for HRP-conjugated antibodies. The sodium azide added as a preservative is an inhibitor of horseradish peroxidase.

How many drops of ReadyProbes 2.5% Normal Goat Serum (1X) should I add to a sample?

Add a sufficient number of drops of ReadyProbes 2.5% Normal Goat Serum (1X) to completely cover the sample.

When would I use goat, horse, or chicken serum for blocking versus FBS or BSA?

For blocking, you should use a serum of the same or closely related species of the secondary antibody being used. If using a secondary antibody that was raised in goat or sheep, use goat serum (Cat. No. R37624 or 31873) for blocking; if using a secondary antibody raised in donkey or horse, use horse serum (Cat. No. R37625 or 31874) for blocking; if using a secondary antibody raised in chicken, use chicken serum (Cat. No. R37626) for blocking. FBS and BSA are alternatives when a specific species of serum is not available.

For Research Use Only. Not for use in diagnostic procedures.