Learn More
Chemglass Inc Reaction Block, 20mL Scintillation Vial, 28mm x 60mm, Black, 17 Position, 24mm Hole Depth
Supplier:  Chemglass Inc CG199101
Encompass
Reaction Block, 20mL Scintillation Vial, 28mm x 60mm, Black, 17 Position, 24mm Hole Depth. Excellent heat transferWon't interfere with magnetic stirringHoles spaced for optimum stirring of all vialsColor-anodized for durability and easy identificationRound reaction blocks are designed to fit directly on 135mm diameter, round top hot plate stirrers. Aluminum construction offers excellent heat transfer without interfering with magnetic stirring. All blocks are anodized for increased strength, durability and chemical resistance. A thermowell is provided for insertion of a temperature sensor or contact thermometer. Available for scintillation vials and dram vials.
Numéro de catalogue. CG199101
Il en reste null