Learn More
New England Biolab Ltd rCutSmart™ Buffer - 5 mls
Numéro de catalogue. 502108568
Encompass
New England Biolabs provides a color-coded 10X NEBuffer with each restriction endonuclease to ensure optimal (100%) activity. Most of our enzymes are supplied with one of four standard NEBuffers. These buffers contain Recombinant Albumin (NEB #B9200). Over 215 restriction enzymes are 100% active in rCutSmart Buffer, making it significantly easier to set up your double digest reactions. Since rCutSmart Buffer includes Recombinant Albumin, there are also fewer tubes and pipetting steps to worry about. Additionally, manyDNA modifying enzymesare 100% active in rCutSmart Buffer, eliminating the need for subsequent purification.
Numéro de catalogue. 502108568 Fournisseur New England Biolab Ltd Code fournisseur B6004S
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
New England Biolabs provides a color-coded 10X NEBuffer with each restriction endonuclease to ensure optimal (100%) activity. Most of our enzymes are supplied with one of four standard NEBuffers. These buffers contain Recombinant Albumin (NEB #B9200). Over 215 restriction enzymes are 100% active in rCutSmart Buffer, making it significantly easier to set up your double digest reactions. Since rCutSmart Buffer includes Recombinant Albumin, there are also fewer tubes and pipetting steps to worry about. Additionally, manyDNA modifying enzymesare 100% active in rCutSmart Buffer, eliminating the need for subsequent purification.