Rat Thymic Stromal Lymphopoietin (TSLP) ELISA Kit for detecting TSLP in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1000pg/ml. Sensitivity: 5.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: A0A0G2JUJ3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.