missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Rat IgG2b, Azide Free, Clone: LTF-2, Negative Control, EMD Millpore™

Numéro de catalogue. MABF1076ZMI
Modifier l'affichage
Click to view available options
Quantité:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
MABF1076ZMI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. MABF1076ZMI Fournisseur MilliporeSigma™ Code fournisseur MABF1076Z

Rat Monoclonal Antibody

Specifically detects IgG2b Clone: LTF-2 in samples, and it is validated for Flow Cytometry

The LTF-2 immunoglobulin is useful as an isotype-matched control. The LTF-2 immunolglobulin has an unknown binding specificity and is used as an isotype control for rat IgG2b antibodies.

Spécifications

Antigène IgG2b
Applications Flow Cytometry
Classification Monoclonal
Clone LTF-2
Formule Purified Rat monoclonal containing 10mM NaH2PO4, 150mM NaCl, pH 7.2. This Monoclonal was purified from tissue culture supernatant via affinity chromatography.
Espèces hôtes Rat
Immunogène Proprietary immunogen
Méthode de purification Affinity Purified
Quantité 100 μg
Statut réglementaire RUO
Disciplines de recherche Inflammation, Immunology
Primaire ou secondaire Primary
Spécifications de test Rat IgG2b Negative Control
Contenu et stockage 2°C to 8°C for one year from date of shipment protected from light; Do not freeze
Isotype IgG2b
Afficher plus de résultats Afficher moins de résultats

For Research Use Only