missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Rat IgG2b, APC, Clone: LTF-2, Negative Control, EMD Millpore™
Description
Specifically detects IgG2b Clone: LTF-2 in samples, and it is validated for Flow Cytometry
The LTF-2 immunoglobulin is useful as an isotype-matched control. The LTF-2 immunolglobulin has an unknown binding specificity and is used as an isotype control for rat IgG2b antibodies.
Spécifications
Spécifications
| Antigen | IgG2b |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | LTF-2 |
| Conjugate | APC |
| Formulation | Purified Rat monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2. This Monoclonal was purified from tissue culture supernatant via affinity chromatography. |
| Host Species | Rat |
| Immunogen | Proprietary immunogen |
| Purification Method | Affinity Purified |
| Quantity | 100 μg |
| Afficher plus de résultats |
For Research Use Only