Learn More
Invitrogen™ Rat anti-Mouse IgG1 Secondary Antibody, eFluor™ 615, eBioscience™
Description
Description: The monoclonal antibody M1-14D12 recognizes Mouse IgG1 antibodies and can be used as a second step reagent in flow cytometry and microscopy. The monoclonal does not recognize other mouse isotypes nor does it crossreact to rat IgG1 or any rat isotype antibodies. Applications Reported: This M1-14D12 antibody has been reported for use in immunohistochemical staining and immunocytochemistry. Applications Tested: This M1-14D12 antibody has been tested by immunocytochemistry and immunohistochemistry after staining with a primary antibody of the mouse IgG1 isotype. This can be used at less than or equal to 5 μg/mL. It is recommended that the antibody be titrated for optimal performance in the assay of interest. This product has not been validated for flow cytometric analysis. Filter Recommendation: When using this eFluor® 615 antibody conjugate, we recommend a filter that will capture the 615 emission wavelength (for example, Excitation 560/55, 585LP, Emission 645/75). A standard Alexa Fluor® 594 filter is acceptable. Excitation: 595 nm; Emission: 615 nm. Filtration: 0.2 μm post-manufacturing filtered.
Spécifications
Spécifications
| Antigène | Mouse IgG1 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Monoclonal |
| Clone | M1-14D12 |
| Concentration | 0.2 mg/mL |
| Conjugué | eFluor 615 |
| Formule | PBS with 0.09% sodium azide; pH 7.2 |
| Espèces hôtes | Rat |
| Méthode de purification | Affinity chromatography |
| Quantité | 100 μg |
| Afficher plus de résultats |