missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rat anti-Mouse IgE Secondary Antibody

Numéro de catalogue. 01672153
Modifier l'affichage
Click to view available options
Quantité:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
01672153 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 01672153 Fournisseur Invitrogen™ Code fournisseur SA135629

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Rat Monoclonal Secondary Antibody

Mouse epsilon heavy chain of immunoglobulin (determined by immunodot). Useful as second antibody in immunoassays, such as ELISA's (good binding on plastic) and immunohistochemistry on frozen sections. Available in unconjugated MON5051, FITC conjugated MON5051F, HRP conjugated MON5050P and Biotin conjugated MON5051B.

Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Spécifications

Antigène Mouse IgE
Applications ELISA, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone LO-ME-3
Concentration 1 mg/mL
Conjugué Unconjugated
Formule PBS with 0.1% sodium azide
Espèces hôtes Rat
Immunogène Proprietary Immunogen.
Méthode de purification Purified
Quantité 1 mL
Statut réglementaire RUO
Primaire ou secondaire Secondary
Espèces cibles Mouse
Contenu et stockage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Type de produit Secondary Antibody
Forme Liquid
Isotype IgG1 κ
Afficher plus de résultats Afficher moins de résultats