missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ RapidTaq qPCR MasterMix (2X)

Catalog No. MP116187100
Change view
Click to view available options
No. of Reactions:
500 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
MP116187100 500 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP116187100 Supplier MP Biomedicals™ Supplier No. 116187100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

RapidTaq qPCR MasterMix (2X) provides rapid extension rates and robust performance.

RapidTaq qPCR MasterMix (2X) contains a ready-to-use next generation Taq polymerase that is engineered for rapid extension rates and robust performance. The 2X MasterMix is convenient and reliable, providing easy set up and reproducible results.

  • Ready-to-Use mastermix
  • Increased sensitivity, yields, and processivity
  • Use with standard or fast cycles of qPCR instruments
  • qPCR products amplified using the RapidTaq DNA polymerase can be used downstream for TA cloning.
  • Reactions run at standard cycles are completed in ∼53 minutes, while reactions run at fast cycles are completed in ∼11 minutes.

Specifications

Content And Storage -20°C
Polymerase RapidTaq (5'-3' polymerase)
Reaction Speed ∼53 min. (Standard)/∼11 min. (Fast)
Exonuclease Activity 5'-3' exonuclease activity, lacks 3'-5' exonuclease activity with 3'-dA-tailed amplicons
For Use With (Application) Real Time PCR (qPCR)
No. of Reactions 500 Reactions
Reaction Format Mastermix
Includes 2X qPCR MasterMix (4 x 1.25 mL), ROX Reference Dye (50 μL)

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.