missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Ranolazine dihydrochloride

Catalog No. 311850
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
311850 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 311850 Supplier Tocris Bioscience™ Supplier No. 3118/50

Antianginal, activates pyruvate dehydrogenase

Antianginal agent with antiarrhythmic properties that acts as a partial fatty acid oxidation inhibitor. Activates pyruvate dehydrogenase in ischemic myocytes to promote glucose oxidation, switching substrate utilization from fatty acids to glucose. Also shown to inhibit late INa and IKr currents.

Chemical Identifiers

CAS 95635-56-6
Molecular Formula C24H35Cl2N3O4

Specifications

Quantity 50 mg
Formula Weight 500.46
Purity >99%
Molecular Weight (g/mol) 500.46
Chemical Name or Material Ranolazine dihydrochloride
Recommended Storage Desiccate at Room Temperature