accessibility menu, dialog, popup

UserName

CAS RN 95635-56-6

IUPAC Name: dihydrogen N-(2,6-dimethylphenyl)-2-{4-[2-hydroxy-3-(2-methoxyphenoxy)propyl]piperazin-1-yl}acetamide dichloride
Synonyms: dihydrogen ranolazine dichloride
Molecular Weight (g/mol): 500.46
Molecular Formula: C24H35Cl2N3O4
InChi Key: RJNSNFZXAZXOFX-UHFFFAOYNA-N
SMILES: [H+].[H+].[Cl-].[Cl-].COC1=CC=CC=C1OCC(O)CN1CCN(CC(=O)NC2=C(C)C=CC=C2C)CC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results