missing translation for 'onlineSavingsMsg'
Learn More

Ranitidine Related Compound C Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015388
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015388 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015388 Supplier Merck Emd Millipore Supplier No. PHR1775100MG

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Pharmaceutical (small molecule)

Specifications

CAS 73851-70-4
Linear Formula C13H22N4O4 S
MDL Number MFCD08063699
Synonym Ranitidine S-oxide; (N-[2-[[[5-[(DIMETHYLAMINO) METHYL]-2-FURANYL]METHYL] SULFINYL]ETHYL]-N-METHYL-2- NITRO-1,1-ETHENEDIAMINE; N-{2-{{{5-[(Dimethylamino)methyl]furan-2-yl}methyl}sulfinyl}ethyl}-N′-methyl-2-nitroethene-1,1-diamine
Type Certified Reference Material
Quantity 100 mg
Formula Weight 330.40
Grade Certified Reference Material