Learn More
RAM Scientific Safet-T-Fil™ Blood Gas Capillaries
100% plastic—no glass to break
$720.71 - $890.57
Specifications
Color Clear
Product Type Blood Gas Tube
Material Plastic
Sterility Non-sterile
View More Specs
Products 4
Catalog Number Mfr. No. Capacity (Metric) Additive Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Additive Price Quantity & Availability  
1491575
View Documents Promotion Details
RAM Scientific
06 0005
115 μL Sodium Heparin
Pack of 250 Each for $722.50
Case of 4 Pack for $2,890.00
Only null left
1491576
View Documents Promotion Details
RAM Scientific
06 0007
150 μL Sodium Heparin
Pack of 250 Each for $890.57
Case of 4 Pack for $3,562.28
Only null left
1491572
View Documents Promotion Details
RAM Scientific
06 0185
115 μL Balanced Heparin
Pack of 250 Each for $720.71
Case of 4 Pack for $2,882.84
Only null left
1491577
View Documents Promotion Details
RAM Scientific
06 0009
230 μL Sodium Heparin
Pack of 250 Each for $811.46
Case of 4 Pack for $3,245.84
Only null left
Description
Description
Blood samples are drawn into the SAFE-T-FILL™ Plastic Capillary Blood Gas Tubes through the same capillary action as glass without the risk of breakage. Capillary tubes are available untreated or heparinized.
  • 100% plastic tubes collect microliter volume capillary blood samples from finger and heel sticks
  • Replaces glass Natelson, Caraway, or mylar-coated glass blood gas capillary tubes
  •  Blood samples are drawn into the capillary tube through the same capillary action as glass without the risk of breakage
  • Capillary tubes are available untreated or heparinized
  • A complete end-to-end fill of the capillary tube accurately measures the stated fill volume
  • Optional end caps minimize the risk of blood spillage and allow for easier handling
  • Optional mixing fleas and magnetic rings (mixing components) allow for uniform mixing to prevent clotting
  • Packaging labels are marked with Lot Number and Expiration Date
Specifications
Specifications
Clear
Blood Gas Tube
Plastic
Non-sterile