missing translation for 'onlineSavingsMsg'
Learn More

RAM Scientific Safet-T-Fil™ Blood Gas Capillaries

Catalog No. 1491575
Change view
Click to view available options
Capacity (Metric):
115 μL
230 μL
Additive:
Balanced Heparin
Sodium Heparin
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Additive
1491575 115 μL Sodium Heparin
1491572 115 μL Balanced Heparin
1491577 230 μL Sodium Heparin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 1491575 Supplier RAM Scientific Supplier No. 06 0005

100% plastic—no glass to break

Blood samples are drawn into the SAFE-T-FILL™ Plastic Capillary Blood Gas Tubes through the same capillary action as glass without the risk of breakage. Capillary tubes are available untreated or heparinized.

  • 100% plastic tubes collect microliter volume capillary blood samples from finger and heel sticks
  • Replaces glass Natelson, Caraway, or mylar-coated glass blood gas capillary tubes
  • Blood samples are drawn into the capillary tube through the same capillary action as glass without the risk of breakage
  • Capillary tubes are available untreated or heparinized
  • A complete end-to-end fill of the capillary tube accurately measures the stated fill volume
  • Optional end caps minimize the risk of blood spillage and allow for easier handling
  • Optional mixing fleas and magnetic rings (mixing components) allow for uniform mixing to prevent clotting
  • Packaging labels are marked with Lot Number and Expiration Date

Recommended Storage

Store at room temperature. Keep dry. Keep away from sunlight.

Specifications

Color Clear
Product Type Blood Gas Tube
Additive Sodium Heparin
Material Plastic
Sterility Non-sterile
Capacity (Metric) 115 μL
For Use With (Application) Intended for blood gas analysis, tests for multiple analytes, or as a transfer device for PKU and POC testing and coated with Sodium Heparin anticoagulant to prevent clotting.