Learn More
RAM Scientific Safet-T-Fil™ Blood Gas Capillaries
100% plastic—no glass to break
Supplier: RAM Scientific 06 0007
Description
Blood samples are drawn into the SAFE-T-FILL™ Plastic Capillary Blood Gas Tubes through the same capillary action as glass without the risk of breakage. Capillary tubes are available untreated or heparinized.
- 100% plastic tubes collect microliter volume capillary blood samples from finger and heel sticks
- Replaces glass Natelson, Caraway, or mylar-coated glass blood gas capillary tubes
- Blood samples are drawn into the capillary tube through the same capillary action as glass without the risk of breakage
- Capillary tubes are available untreated or heparinized
- A complete end-to-end fill of the capillary tube accurately measures the stated fill volume
- Optional end caps minimize the risk of blood spillage and allow for easier handling
- Optional mixing fleas and magnetic rings (mixing components) allow for uniform mixing to prevent clotting
- Packaging labels are marked with Lot Number and Expiration Date
Recommended Storage
Store at room temperature. Keep dry. Keep away from sunlight.
Chemical Identifiers
| Clear | |
| Blood Gas Tube | |
| Plastic | |
| Non-sterile |
Specifications
| Clear | |
| Sodium Heparin | |
| Non-sterile | |
| Intended for blood gas analysis, tests for multiple analytes, or as a transfer device for PKU and POC testing and coated with Sodium Heparin anticoagulant to prevent clotting. |
| Blood Gas Tube | |
| Plastic | |
| 150 μL |