missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Raloxifene hydrochloride

Catalog No. AC466500010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466500010 1 g
AC466500050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC466500010 Supplier Thermo Scientific Chemicals Supplier No. 466500010

CAS: 82640-04-8 | C28H28ClNO4S, C28H27NO4S·HCl | 510.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 82640-04-8
Molecular Formula C28H27NO4S·HCl

Specifications

Quantity 1 g
Solubility Solubility aq. soln.: .. Other solubilities: soluble in dmso
Melting Point 263.0°C
Formula Weight 510.04
InChI Key BKXVVCILCIUCLG-UHFFFAOYSA-N
SMILES [H+].[Cl-].OC1=CC=C(C=C1)C1=C(C(=O)C2=CC=C(OCCN3CCCCC3)C=C2)C2=CC=C(O)C=C2S1
IUPAC Name hydrogen 2-(4-hydroxyphenyl)-3-{4-[2-(piperidin-1-yl)ethoxy]benzoyl}-1-benzothiophen-6-ol chloride
Molecular Weight (g/mol) 510.05
Chemical Name or Material Raloxifene hydrochloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Suspected of causing cancer.

May damage fertility or the unborn child.

Precautionary Statement GHS P Statement Obtain special instructions before use.

Wear protective gloves/protective clothing/eye protection/face protection.

IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only