Learn More
MilliporeSigma™ Supelco™ radiello™ BTEX/VOC Starter Kit, CS2 Desorption
Catalog No. 111010643
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
111010643 1 Kit
1 options
Catalog No. 111010643 Supplier MilliporeSigma™ Supelco™ Supplier No. SGMARAD130S
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
radiello™ Starter Kits are ideal for introducing you to the product and include everything you need to take one complete sample. No additional parts are required.
Each RAD130S Starter Kit includes: Two radiello™ cartridge adsorbents (RAD130) for VOCs - CS2 Desorption (Note: 1 x cartridge for sampling; and 1 x for blank), 1 x Triangular support plate (RAD121), 1 x vertical adapter for personal sampling (RAD122), 1 x white diffusive body (RAD120) and Detailed sampling/analysis instructions
Specifications
Includes 2 RAD130 adsorbing cartridges and barcode labels, 1 RAD120 white diffusive body, 1 RAD121 triangular support plate, 1 instruction sheet
Quantity 1 Kit
radiello is a trademark of Institi Clinici Scientifici Maugeri