Learn More
Alkali Scientific Radiant™ qPCR Genotyping Mix, No-ROX, Suitable for Dual-labeled Taqman™ and Other Probes, 100 Reactions, (1 X 1mL)
Catalog No. 502389453
Encompass
Using the latest advancements in polymerase technology and advanced buffer chemistry Radiant™ Genotyping Mix No-Rox offers market leading performance with minimal optimization. Radiant™ Genotyping Mix No-Rox is a kit designed for use in dual-labeled probe based genotyping assays including Taqman, Molecular Beacon and Scorpion probe genotyping. Radiant™ GenotypingMix is fully compatible with ABI TaqMan Pre-Designed SNP Genotyping assays. Radiant™ Genotyping Mix uses proprietary small molecular inhibitor technology that prevents formation of primer-dimers to improve reaction sensitivity and specificity. Highthroughput screening has resulted in a buffer system that allows efficient amplification from GC-rich and AT-rich templates, under fast and standard cycling conditions.
Catalog No. 502389453 Supplier Alkali Scientific Supplier No. QG3001
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Using the latest advancements in polymerase technology and advanced buffer chemistry Radiant™ Genotyping Mix No-Rox offers market leading performance with minimal optimization. Radiant™ Genotyping Mix No-Rox is a kit designed for use in dual-labeled probe based genotyping assays including Taqman, Molecular Beacon and Scorpion probe genotyping. Radiant™ GenotypingMix is fully compatible with ABI TaqMan Pre-Designed SNP Genotyping assays. Radiant™ Genotyping Mix uses proprietary small molecular inhibitor technology that prevents formation of primer-dimers to improve reaction sensitivity and specificity. Highthroughput screening has resulted in a buffer system that allows efficient amplification from GC-rich and AT-rich templates, under fast and standard cycling conditions.