Learn More
Alkali Scientific Radiant™ Probe 1-Step No-ROX qPCR Kit, 100 Reactions, (1 X 1mL 2X Mix, 1 X 0.2mL 20X RTase)
Numéro de catalogue. 502389429
Encompass
Alkali Scientific offers Radiant™ Probe 1-Step No-ROX qPCR Kits, a high-performance portfolio of 2X ready-to-use real-time PCR reagents designed for best-in- class quantitative PCR. Radiant™ qPCR Kits are engineered for robust real-time PCR with earlier quantification cycle values (Ct), industry-leading sensitivity (increased limit of detection) and exceptional speed (rapid extension rates). The proprietary buffer system allows for highly efficient amplification of GC-rich and AT-rich sequences in addition to dramatic improvements in PCR sensitivity in low- copy assays and PCR conditions conducive to ultra-fast amplification. The Radiant™ Probe 1-Step No-ROX qPCR Kit utilizes a proprietary, Green non-inhibitory intercalating dye for robust, fluorescent detection.
Numéro de catalogue. 502389429 Fournisseur Alkali Scientific Code fournisseur QN6001
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Alkali Scientific offers Radiant™ Probe 1-Step No-ROX qPCR Kits, a high-performance portfolio of 2X ready-to-use real-time PCR reagents designed for best-in- class quantitative PCR. Radiant™ qPCR Kits are engineered for robust real-time PCR with earlier quantification cycle values (Ct), industry-leading sensitivity (increased limit of detection) and exceptional speed (rapid extension rates). The proprietary buffer system allows for highly efficient amplification of GC-rich and AT-rich sequences in addition to dramatic improvements in PCR sensitivity in low- copy assays and PCR conditions conducive to ultra-fast amplification. The Radiant™ Probe 1-Step No-ROX qPCR Kit utilizes a proprietary, Green non-inhibitory intercalating dye for robust, fluorescent detection.