Learn More
Alkali Scientific Radiant™ dNTP Mix, 100mM each, (4 x 4 x 1mL vial each nucleotide), total 16.0mL
Numéro de catalogue. 503026490
Encompass
Radiant dNTP Mix contains premixed aqueous solutions of dATP, dCTP, dGTP and dTTP The Radiant dNTP mix is ultra pure (more than 99%), stable after multiple freeze-thaw cycles and perfect for a wide variety of applications. Ultra pure Stable Versatile
Numéro de catalogue. 503026490 Fournisseur Alkali Scientific Code fournisseur C769
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Radiant dNTP Mix contains premixed aqueous solutions of dATP, dCTP, dGTP and dTTP The Radiant dNTP mix is ultra pure (more than 99%), stable after multiple freeze-thaw cycles and perfect for a wide variety of applications. Ultra pure Stable Versatile