missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Raclopride | |
| 54°C | |
| MFCD02179386 | |
| raclopride, s-3,5-dichloro-n-1-ethylpyrrolidin-2-yl methyl-2-hydroxy-6-methoxybenzamide, 3h raclopride, 3h-raclopride, unii-430k3soz7g, chembl8809, raclopridum latin, racloprida spanish, raclopride inn:ban, dsstox_cid_25687 | |
| CCN1CCCC1CNC(=O)C2=C(C(=CC(=C2OC)Cl)Cl)O | |
| 347.236 | |
| 347.24 | |
| Crystalline Powder |
| 84225-95-6 | |
| C15H20Cl2N2O3 | |
| 1 g | |
| WAOQONBSWFLFPE-VIFPVBQESA-N | |
| 3,5-dichloro-N-[[(2S)-1-ethylpyrrolidin-2-yl]methyl]-2-hydroxy-6-methoxybenzamide | |
| 3033769 | |
| ≥98.0% (HPLC,N) |
Chemical Identifiers
| 84225-95-6 | |
| 347.236 | |
| WAOQONBSWFLFPE-VIFPVBQESA-N | |
| 3033769 | |
| CCN1CCCC1CNC(=O)C2=C(C(=CC(=C2OC)Cl)Cl)O |
| C15H20Cl2N2O3 | |
| MFCD02179386 | |
| raclopride, s-3,5-dichloro-n-1-ethylpyrrolidin-2-yl methyl-2-hydroxy-6-methoxybenzamide, 3h raclopride, 3h-raclopride, unii-430k3soz7g, chembl8809, raclopridum latin, racloprida spanish, raclopride inn:ban, dsstox_cid_25687 | |
| 3,5-dichloro-N-[[(2S)-1-ethylpyrrolidin-2-yl]methyl]-2-hydroxy-6-methoxybenzamide |
Safety and Handling
RTECSNumber : CV3463500
TSCA : No