missing translation for 'onlineSavingsMsg'
Learn More

Raclopride 98.0+%, TCI America™

Catalog No. p-7135692
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R0094200MG 200 mg
R00941G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R0094200MG Supplier TCI America Supplier No. R0094200MG

Chemical Identifiers

CAS 84225-95-6
Molecular Formula C15H20Cl2N2O3
Molecular Weight (g/mol) 347.236
MDL Number MFCD02179386
InChI Key WAOQONBSWFLFPE-VIFPVBQESA-N
Synonym raclopride, s-3,5-dichloro-n-1-ethylpyrrolidin-2-yl methyl-2-hydroxy-6-methoxybenzamide, 3h raclopride, 3h-raclopride, unii-430k3soz7g, chembl8809, raclopridum latin, racloprida spanish, raclopride inn:ban, dsstox_cid_25687
PubChem CID 3033769
IUPAC Name 3,5-dichloro-N-[[(2S)-1-ethylpyrrolidin-2-yl]methyl]-2-hydroxy-6-methoxybenzamide
SMILES CCN1CCCC1CNC(=O)C2=C(C(=CC(=C2OC)Cl)Cl)O

Specifications

Melting Point 54°C
Quantity 200 mg
Formula Weight 347.24
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Raclopride
RTECSNumber CV3463500
TSCA No