accessibility menu, dialog, popup

UserName

CH3N(R)NHOHOClClOCH3

CAS RN 84225-95-6

IUPAC Name: 3,5-dichloro-N-{[(2R)-1-ethylpyrrolidin-2-yl]methyl}-2-hydroxy-6-methoxybenzamide
Synonyms: raclopride
Molecular Weight (g/mol): 347.24
Molecular Formula: C15H20Cl2N2O3
InChi Key: WAOQONBSWFLFPE-SECBINFHSA-N
SMILES: CCN1CCC[C@@H]1CNC(=O)C1=C(O)C(Cl)=CC(Cl)=C1OC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results