missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Racepinephrine Hydrochloride, USP, 97-102%, Spectrum™ Chemical
E1418, 329-63-5, C9H13NO3·HCl
Supplier: Spectrum Chemical Mfg Cor E14181KGBL
Description
Spectrum™ Chemical Racepinephrine Hydrochloride, USP is a bronchodilator used to relieve occasional symptoms of asthma, such as wheezing, chest tightness, and feeling short of breath. All Spectrum Chemical USP products, including all USP hydrochloride products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 329-63-5 | |
| 0.005 | |
| C9H14ClNO3 | |
| -1° to +1° | |
| [H+].[Cl-].CNCC(O)C1=CC=C(O)C(O)=C1 | |
| 219.67 | |
| USP |
| 1 | |
| Amber Glass Bottle | |
| 1 kg | |
| ATADHKWKHYVBTJ-UHFFFAOYNA-N | |
| hydrogen 4-[1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol chloride | |
| 97 to 102% | |
| Crystalline Powder or Lumps |
Chemical Identifiers
| 329-63-5 | |
| 219.67 | |
| hydrogen 4-[1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol chloride |
| C9H14ClNO3 | |
| ATADHKWKHYVBTJ-UHFFFAOYNA-N | |
| [H+].[Cl-].CNCC(O)C1=CC=C(O)C(O)=C1 |