accessibility menu, dialog, popup

UserName

HClH3CNHOHOHHO

CAS RN 329-63-5

IUPAC Name: hydrogen 4-[1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol chloride
Synonyms: hydrogen (+-)-adrenaline chloride
Molecular Weight (g/mol): 219.67
Molecular Formula: C9H14ClNO3
InChi Key: ATADHKWKHYVBTJ-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CNCC(O)C1=CC=C(O)C(O)=C1
Molecular Weight (g/mol) 
  • (2)
  • (6)
Percent Purity 
  • (2)
  • (6)
Physical Form 
  • (2)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Grade 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (6)
  • (6)
  • (2)
  • (2)
  • (6)
  • (2)
  • (6)
  • (2)
  • (6)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (6)